MilliporeSigma
  • Sustained Secretion of the Antimicrobial Peptide S100A7 Is Dependent on the Downregulation of Caspase-8.
Sustained Secretion of the Antimicrobial Peptide S100A7 Is Dependent on the Downregulation of Caspase-8.
Cell reports (2019-11-28)
Tanay Bhatt, Aishwarya Bhosale, Bhavya Bajantri, Mruthyunjaya Swamy Mathapathi, Abrar Rizvi, Giorgio Scita, Amitabha Majumdar, Colin Jamora
ABSTRACT
Antimicrobial peptides (AMPs) are the body's natural innate immune defense against a spectrum of pathogens and can also modulate cell proliferation, chemotaxis, angiogenesis, wound healing, and immune cell activity. Harnessing these diverse functions for prophylactic use is contingent upon understanding the regulatory mechanisms governing their unconventional secretion from cells. Analysis of the secretion of S100A7 (Psoriasin), an abundant AMP stored in differentiated keratinocytes of the skin, has revealed an unexpected biphasic secretory response to bacterial exposure. The core components regulating S100A7 secretion are NFκB/p38MAPK, caspase-1, and interleukin (IL)-1α. The initial activation of this core machinery is mediated by Toll-like receptor signaling, whereas the chronic response is mediated by Caspase-8 downregulation. Interestingly, there is a concomitant downregulation of Caspase-8 in inflammatory skin diseases wherein S100A7 is constitutively released. These results highlight the potential of targeting these components to control the release of AMPs from the skin in both homeostatic and disease conditions.
Sigma-Aldrich
Potassium nitrate, puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
MISSION® esiRNA, targeting human CASP8
Sigma-Aldrich
MISSION® esiRNA, targeting human TLR5