MilliporeSigma
  • PCDH17 increases the sensitivity of colorectal cancer to 5-fluorouracil treatment by inducing apoptosis and autophagic cell death.
PCDH17 increases the sensitivity of colorectal cancer to 5-fluorouracil treatment by inducing apoptosis and autophagic cell death.
Signal transduction and targeted therapy (2019-12-10)
Shuiping Liu, Haoming Lin, Da Wang, Qiang Li, Hong Luo, Guoxiong Li, Xiaohui Chen, Yongqiang Li, Peng Chen, Bingtao Zhai, Wengang Wang, Ruonan Zhang, Bi Chen, Mingming Zhang, Xuemeng Han, Qiujie Li, Liuxi Chen, Ying Liu, Xiaying Chen, Guohua Li, Yu Xiang, Ting Duan, Jiao Feng, Jianshu Lou, Xingxing Huang, Qin Zhang, Ting Pan, Lili Yan, Ting Jin, Wenzheng Zhang, Lvjia Zhuo, Yitian Sun, Tian Xie, Xinbing Sui
ABSTRACT
5-Fluorouracil (5-FU) is known as a first-line chemotherapeutic agent against colorectal cancer (CRC), but drug resistance occurs frequently and significantly limits its clinical success. Our previous study showed that the protocadherin 17 (PCDH17) gene was frequently methylated and functioned as a tumor suppressor in CRC. However, the relationship between PCDH17 and 5-FU resistance in CRC remains unclear. Here, we revealed that PCDH17 was more highly expressed in 5-FU-sensitive CRC tissues than in 5-FU-resistant CRC tissues, and high expression of PCDH17 was correlated with high BECN1 expression. Moreover, this expression profile contributed to superior prognosis and increased survival in CRC patients. Restoring PCDH17 expression augmented the 5-FU sensitivity of CRC in vitro and in vivo by promoting apoptosis and autophagic cell death. Furthermore, autophagy played a dominant role in PCDH17-induced cell death, as an autophagy inhibitor blocked cell death to a greater extent than the pancaspase inhibitor Z-VAD-FMK. PCDH17 inhibition by siRNA decreased the autophagy response and 5-FU sensitivity. Mechanistically, we showed that c-Jun NH2-terminal kinase (JNK) activation was a key determinant in PCDH17-induced autophagy. The compound SP600125, an inhibitor of JNK, suppressed autophagy and 5-FU-induced cell death in PCDH17-reexpressing CRC cells. Taken together, our findings suggest for the first time that PCDH17 increases the sensitivity of CRC to 5-FU treatment by inducing apoptosis and JNK-dependent autophagic cell death. PCDH17 may be a potential prognostic marker for predicting 5-FU sensitivity in CRC patients.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Z-Val-Ala-Asp fluoromethyl ketone, powder, ≥90% (TLC)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Anti-PCDH17 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Necrostatin-1, ≥98% (HPLC)
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)