MilliporeSigma
  • Novel enzymatic single-nucleotide modification of DNA oligomer: prevention of incessant incorporation of nucleotidyl transferase by ribonucleotide-borate complex.
Novel enzymatic single-nucleotide modification of DNA oligomer: prevention of incessant incorporation of nucleotidyl transferase by ribonucleotide-borate complex.
Nucleic acids research (2019-07-19)
Eui Kyoung Jang, Ryeo Gang Son, Seung Pil Pack
ABSTRACT
Terminal deoxynucleotidyl transferase (TdT), which mediates template-independent polymerization with low specificity for nucleotides, has been used for nucleotide extension of DNA oligomers. One concern is that it is difficult to control the number of incorporated nucleotides, which is a limitation on the use of TdT for single-nucleotide incorporation of DNA oligomers. Herein, we uncovered an interesting inhibitory effect on TdT when ribonucleotide substrates (rNTPs) were employed in a borate buffer. On the basis of unique inhibitory effects of the ribonucleotide-borate complex, we developed a novel enzymatic method for single-nucleotide incorporation of a DNA oligomer with a modified rNTP by TdT. Single-nucleotide incorporation of a DNA oligomer with various modified rNTPs containing an oxanine, biotin, aminoallyl or N6-propargyl group was achieved with a high yield. The 'TdT in rNTP-borate' method is quite simple and efficient for preparing a single-nucleotide modified DNA oligomer, which is useful for the design of functional DNA-based systems.
Sigma-Aldrich
Guanosine 5′-triphosphate sodium salt hydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
N6-Propargyl-ATP sodium salt
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of 30 mg
Sigma-Aldrich
Uridine 5′-triphosphate trisodium salt hydrate, from yeast, Type III, ≥96%
Sigma-Aldrich
Aminoallyl-UTP sodium salt, ≥75%, lyophilized powder