MilliporeSigma
  • Inhibition of the Formation In Vitro of Putatively Carcinogenic Metabolites Derived from S. haematobium and O. viverrini by Combination of Drugs with Antioxidants.
Inhibition of the Formation In Vitro of Putatively Carcinogenic Metabolites Derived from S. haematobium and O. viverrini by Combination of Drugs with Antioxidants.
Molecules (Basel, Switzerland) (2019-11-17)
Maria João Gouveia, Verónica Nogueira, Bruno Araújo, Fátima Gärtner, Nuno Vale
ABSTRACT
Infections caused by Schistosoma haematobium and Opisthorchis viverrini are classified as carcinogenic. Although carcinogenesis might be a multifactorial process, it has been postulated that these helminth produce/excrete oxysterols and estrogen-like metabolites that might act as initiators of their infection-associated carcinogenesis. Current treatment and control of these infections rely on a single drug, praziquantel, that mainly targets the parasites and not the pathologies related to the infection including cancer. Thus, there is a need to search for novel therapeutic alternatives that might include combinations of drugs and drug repurposing. Based on these concepts, we propose a novel therapeutic strategy that combines drugs with molecule antioxidants. We evaluate the efficacy of a novel therapeutic strategy to prevent the formation of putative carcinogenic metabolites precursors and DNA adducts. Firstly, we used a methodology previously established to synthesize metabolites precursors and DNA adducts in the presence of CYP450. Then, we evaluated the inhibition of their formation induced by drugs and antioxidants alone or in combination. Drugs and resveratrol alone did not show a significant inhibitory effect while N-acetylcysteine inhibited the formation of most metabolite precursors and DNA adducts. Moreover, the combinations of classical drugs with antioxidants were more effective rather than compounds alone. This strategy might be a valuable tool to prevent the initiation of helminth infection-associated carcinogenesis.
Sigma-Aldrich
CypExpress 2E1 Cytochrome P450 human
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from calf thymus, Type I, fibers
Sigma-Aldrich
Glycocholic acid hydrate, synthetic, ≥97% (HPLC)
Sigma-Aldrich
CypExpress 3A4 Cytochrome P450 human
Sigma-Aldrich
CypExpress 1A1 Cytochrome P450 human