MilliporeSigma
  • Quantitative proteomic analyses reveal that GPX4 downregulation during myocardial infarction contributes to ferroptosis in cardiomyocytes.
Quantitative proteomic analyses reveal that GPX4 downregulation during myocardial infarction contributes to ferroptosis in cardiomyocytes.
Cell death & disease (2019-11-07)
Tae-Jun Park, Jei Hyoung Park, Ga Seul Lee, Ji-Yoon Lee, Ji Hye Shin, Min Wook Kim, Yong Sook Kim, Jeong-Yoon Kim, Kyoung-Jin Oh, Baek-Soo Han, Won-Kon Kim, Youngkeun Ahn, Jeong Hee Moon, Jaewhan Song, Kwang-Hee Bae, Do Han Kim, Eun-Woo Lee, Sang Chul Lee
ABSTRACT
Ischaemic heart disease (IHD) is the leading cause of death worldwide. Although myocardial cell death plays a significant role in myocardial infarction (MI), its underlying mechanism remains to be elucidated. To understand the progression of MI and identify potential therapeutic targets, we performed tandem mass tag (TMT)-based quantitative proteomic analysis using an MI mouse model. Gene ontology (GO) analysis and gene set enrichment analysis (GSEA) revealed that the glutathione metabolic pathway and reactive oxygen species (ROS) pathway were significantly downregulated during MI. In particular, glutathione peroxidase 4 (GPX4), which protects cells from ferroptosis (an iron-dependent programme of regulated necrosis), was downregulated in the early and middle stages of MI. RNA-seq and qRT-PCR analyses suggested that GPX4 downregulation occurred at the transcriptional level. Depletion or inhibition of GPX4 using specific siRNA or the chemical inhibitor RSL3, respectively, resulted in the accumulation of lipid peroxide, leading to cell death by ferroptosis in H9c2 cardiomyoblasts. Although neonatal rat ventricular myocytes (NRVMs) were less sensitive to GPX4 inhibition than H9c2 cells, NRVMs rapidly underwent ferroptosis in response to GPX4 inhibition under cysteine deprivation. Our study suggests that downregulation of GPX4 during MI contributes to ferroptotic cell death in cardiomyocytes upon metabolic stress such as cysteine deprivation.
Sigma-Aldrich
MISSION® esiRNA, targeting human GPX4
Sigma-Aldrich
Liproxstatin-1, >98% (HPLC)
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Dimethyl malonate, 98%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid