MilliporeSigma
  • A reporter mouse for non-invasive detection of toll-like receptor ligands induced acute phase responses.
A reporter mouse for non-invasive detection of toll-like receptor ligands induced acute phase responses.
Scientific reports (2019-12-15)
Chun-Fang Huang, Shang-Yi Chiu, Hung-Wen Huang, Bing-Ho Cheng, Hsiu-Min Pan, Wei-Lun Huang, Hsiao-Hui Chang, Chia-Chi Liao, Si-Tse Jiang, Yu-Chia Su
ABSTRACT
The acute phase response (APR) is a systemic first-line defense against challenges including infection, trauma, stress, and neoplasia. Alteration of acute phase protein (APP) levels in plasma is the most important change during acute phase response. C-reactive protein (CRP), which increases dramatically during inflammation onset, is an indicator of inflammation. To monitor the process of APR, we generated human CRP promoter-driven luciferase transgenic (hCRP-Luc) mice to quantify the hCRP promoter activation in vivo. The naïve female hCRP-Luc mice express low basal levels of liver bioluminescence, but the naïve male hCRP-Luc mice do not. Thus, female hCRP-Luc mice are suitable for monitoring the process of APR. The liver bioluminescence of female hCRP-Luc mice can be induced by several toll-like receptor (TLR) ligands. The expression of liver bioluminescence was highly sensitive to endotoxin stimulation in a dose-dependent manner. On-off-on bioluminescence response was noted in female hCRP-Luc mice upon two endotoxin stimulations one month apart. The LPS-induced bioluminescence of the female hCRP-Luc mice was IL-6-mediated and associated with APP alpha-1-acid glycoprotein expression. In conclusion, the female hCRP-Luc mouse is a non-invasive, sensitive and reusable reporter tool for APR.
Sigma-Aldrich
E-Toxate Endotoxin standard, for endotoxin quantitation
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Zymosan A from Saccharomyces cerevisiae, for inducing inflamatory response