MilliporeSigma
  • Mitochondrial dysfunction increases fatty acid β-oxidation and translates into impaired neuroblast maturation.
Mitochondrial dysfunction increases fatty acid β-oxidation and translates into impaired neuroblast maturation.
FEBS letters (2019-08-23)
Matteo Audano, Silvia Pedretti, Maurizio Crestani, Donatella Caruso, Emma De Fabiani, Nico Mitro
ABSTRACT
The metabolic transition from anaerobic glycolysis and fatty acid β-oxidation to glycolysis coupled to oxidative phosphorylation is a key process for the transition of quiescent neural stem cells to proliferative neural progenitor cells. However, a full characterization of the metabolic shift and the involvement of mitochondria during the last step of neurogenesis, from neuroblasts to neuron maturation, is still elusive. Here, we describe a model of neuroblasts, Neuro2a cells, with impaired differentiation capacity due to mitochondrial dysfunction. Using a detailed biochemical characterization consisting of steady-state metabolomics and metabolic flux analysis, we find increased fatty acid β-oxidation as a peculiar feature of neuroblasts with altered mitochondria. The consequent metabolic switch favors neuroblast proliferation at the expense of neuron maturation.
Sigma-Aldrich
L-Glutamine-13C5, 98 atom % 13C, 95% (CP)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Palmitic acid-13C16, 99 atom % 13C, 99% (CP)
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
D-Glucose-13C6, ≥99 atom % 13C, ≥99% (CP)