MilliporeSigma
  • Dissociating stable nitrogen molecules under mild conditions by cyclic strain engineering.
Dissociating stable nitrogen molecules under mild conditions by cyclic strain engineering.
Science advances (2019-11-09)
Gao-Feng Han, Xiang-Mei Shi, Seok-Jin Kim, Jeonghun Kim, Jong-Pil Jeon, Hyuk-Jun Noh, Yoon-Kwang Im, Feng Li, Young Rang Uhm, Chul Sung Kim, Qing Jiang, Jong-Beom Baek
ABSTRACT
All quiet on the nitrogen front. The dissociation of stable diatomic nitrogen molecules (N2) is one of the most challenging tasks in the scientific community and currently requires both high pressure and high temperature. Here, we demonstrate that N2 can be dissociated under mild conditions by cyclic strain engineering. The method can be performed at a critical reaction pressure of less than 1 bar, and the temperature of the reaction container is only 40°C. When graphite was used as a dissociated N* receptor, the normalized loading of N to C reached as high as 16.3 at/at %. Such efficient nitrogen dissociation is induced by the cyclic loading and unloading mechanical strain, which has the effect of altering the binding energy of N, facilitating adsorption in the strain-free stage and desorption in the compressive strain stage. Our finding may lead to opportunities for the direct synthesis of N-containing compounds from N2.
Sigma-Aldrich
Sodium sulfide
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)