MilliporeSigma
  • DNA-Dependent Protein Kinase Drives Prostate Cancer Progression through Transcriptional Regulation of the Wnt Signaling Pathway.
DNA-Dependent Protein Kinase Drives Prostate Cancer Progression through Transcriptional Regulation of the Wnt Signaling Pathway.
Clinical cancer research : an official journal of the American Association for Cancer Research (2019-07-04)
Vishal Kothari, Jonathan F Goodwin, Shuang G Zhao, Justin M Drake, Yi Yin, S Laura Chang, Joseph R Evans, Kari Wilder-Romans, Kristina Gabbara, Emanuela Dylgjeri, Jonathan Chou, Grace Sun, Scott A Tomlins, Rohit Mehra, Kristen Hege, Ellen H Filvaroff, Edward M Schaeffer, R Jeffrey Karnes, David A Quigley, Dana E Rathkopf, Housheng H He, Corey Speers, Daniel E Spratt, Luke A Gilbert, Alan Ashworth, Arul M Chinnaiyan, Ganesh V Raj, Karen E Knudsen, Felix Y Feng
ABSTRACT
Protein kinases are known to play a prominent role in oncogenic progression across multiple cancer subtypes, yet their role in prostate cancer progression remains underexplored. The purpose of this study was to identify kinases that drive prostate cancer progression.Experimental Design: To discover kinases that drive prostate cancer progression, we investigated the association between gene expression of all known kinases and long-term clinical outcomes in tumor samples from 545 patients with high-risk disease. We evaluated the impact of genetic and pharmacologic inhibition of the most significant kinase associated with metastatic progression in vitro and in vivo. DNA-dependent protein kinase (DNAPK) was identified as the most significant kinase associated with metastatic progression in high-risk prostate cancer. Inhibition of DNAPK suppressed the growth of both AR-dependent and AR-independent prostate cancer cells. Gene set enrichment analysis nominated Wnt as the top pathway associated with DNAPK. We found that DNAPK interacts with the Wnt transcription factor LEF1 and is critical for LEF1-mediated transcription. Our data show that DNAPK drives prostate cancer progression through transcriptional regulation of Wnt signaling and is an attractive therapeutic target in aggressive prostate cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human PRKDC
Sigma-Aldrich
ChIPAb+ LEF1 - ChIP Validated Antibody and Primer Set, from mouse
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
IWP-2, ≥98% (HPLC)