MilliporeSigma
NMR carbohydrate profile in tracing acacia honey authenticity.
Food chemistry (2019-11-23)
Elisabetta Schievano, Marco Sbrizza, Valentina Zuccato, Lucia Piana, Marco Tessari
ABSTRACT
The sugar profile in honey can be used as a fingerprint to confirm the authenticity or reveal the adulteration of the product by sweetener addition. In this work, we have accurately determined the profile of 20 minor saccharides in a set of 46 European acacia honeys using a recently proposed NMR approach based on the CSSF-TOCSY experiment. Comparison of this reference profile with the sugar composition of several Chinese honey samples of the same declared botanical origin has revealed important differences. A detailed analysis of the saccharide profile of these Chinese honeys suggests product adulteration by overfeeding bee colonies with industrial sugars syrups during the main nectar flow period.
Sigma-Aldrich
Kojibiose, ≥98% (HPLC)
Sigma-Aldrich
Erlose, ≥94% (HPLC)
Sigma-Aldrich
β-Gentiobiose, ≥85% (remainder primarily α-anomer)
Sigma-Aldrich
Palatinose hydrate, ≥99% (GC)
Supelco
1-Kestose, analytical standard
Sigma-Aldrich
Isomaltose, ~98% (TLC)
Sigma-Aldrich
Melibiose, ≥98% (HPLC)
Sigma-Aldrich
L-Rhamnose monohydrate, ≥99%