MilliporeSigma
  • Industrial Riboflavin Fermentation Broths Represent a Diverse Source of Natural Saturated and Unsaturated Lactones.
Industrial Riboflavin Fermentation Broths Represent a Diverse Source of Natural Saturated and Unsaturated Lactones.
Journal of agricultural and food chemistry (2019-04-19)
Florian Birk, Marco A Fraatz, Patrick Esch, Sven Heiles, Ralf Pelzer, Holger Zorn
ABSTRACT
Fermentation broths of Ashbya gossypii from the industrial production of riboflavin emit an intense floral, fruity, and nutty smell. Typical Ehrlich pathway products, such as 2-phenylethan-1-ol and 2-/3-methylbutan-1-ol, were detected in large amounts as well as some intensely smelling saturated and unsaturated lactones, e.g., γ-decalactone and γ-(Z)-dodec-6-enlactone. An aroma extract dilution analysis identified 2-phenylethan-1-ol and γ-(Z)-dodec-6-enlactone as the main contributors to the overall aroma, with flavor dilution factors of 32 768. The position of the double bonds of unsaturated lactones was determined by the Paternò-Büchi reaction, and reference compounds that were not available commercially were synthesized to elucidate the structures of the uncommon lactones. The absolute configuration and enantiomeric excess values of the lactones were determined by converting the lactones to their corresponding Mosher's esters. In addition, the odor impressions and odor thresholds in air were determined.
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
1-Heptyne, 98%
Sigma-Aldrich
3-Chloroperbenzoic acid, ≤77%
Sigma-Aldrich
Phenylacetyl chloride, 98%
Sigma-Aldrich
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Propylbenzene, 98%
Sigma-Aldrich
N-Chlorosuccinimide, 98%