MilliporeSigma
  • Identification of factors involved in the anti-tumor activity of carnosine on glioblastomas using a proteomics approach.
Identification of factors involved in the anti-tumor activity of carnosine on glioblastomas using a proteomics approach.
Cancer investigation (2011-02-25)
Ansgar Asperger, Christof Renner, Mandy Menzel, Rolf Gebhardt, Jürgen Meixensberger, Frank Gaunitz
ABSTRACT
Human glioblastoma multiforme is the most malignant brain tumor in adults and is difficult to treat. Recently, it was demonstrated that the dipeptide carnosine inhibits tumor growth, but the main molecular targets are not known. Therefore, a proteomics study with glioblastoma cells treated with carnosine was performed. Two-dimensional gel electrophoresis and matrix-assisted laser desorption/ionization time of flight detected 31 proteins expressed differentially under the influence of carnosine. Finally, peptide mass fingerprinting identified the "BCL2-associated athanogene 2" and the "von Hippel-Lindau binding protein 1" among other proteins, linking the action of carnosine to protein folding and HIF-1α signalling.