MilliporeSigma
  • The short-chain perfluorinated compounds PFBS, PFHxS, PFBA and PFHxA, disrupt human mesenchymal stem cell self-renewal and adipogenic differentiation.
The short-chain perfluorinated compounds PFBS, PFHxS, PFBA and PFHxA, disrupt human mesenchymal stem cell self-renewal and adipogenic differentiation.
Journal of environmental sciences (China) (2019-12-22)
Shuyu Liu, Renjun Yang, Nuoya Yin, Francesco Faiola
ABSTRACT
Per- and polyfluorinated alkyl substances (PFASs) are commonly used in industrial processes and daily life products. Because they are persistent, they accumulate in the environment, wildlife and humans. Although many studies have focused on two of the most representative PFASs, PFOS and PFOA, the potential toxicity of short-chain PFASs has not yet been given sufficient attention. We used a battery of assays to evaluate the toxicity of several four-carbon and six-carbon perfluorinated sulfonates and carboxyl acids (PFBS, PFHxS, PFBA and PFHxA), with a human mesenchymal stem cell (hMSC) system. Our results demonstrate significant cyto- and potential developmental toxicity for all the compounds analyzed, with shared but also distinct mechanisms of toxicity. Moreover, the effects of PFBS and PFHxS were stronger than those of PFBA and PFHxA, but occurred at higher doses compared to PFOS or PFOA.
Sigma-Aldrich
Fura 2-AM, ≥95% (HPLC)
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, ≥95%
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Undecafluorohexanoic acid, ≥97.0% (T)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Nile Red, Technical grade
Sigma-Aldrich
Potassium nonafluoro-1-butanesulfonate, 98%
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)