MilliporeSigma
  • Carboxylated branched poly(β-amino ester) nanoparticles enable robust cytosolic protein delivery and CRISPR-Cas9 gene editing.
Carboxylated branched poly(β-amino ester) nanoparticles enable robust cytosolic protein delivery and CRISPR-Cas9 gene editing.
Science advances (2019-12-17)
Yuan Rui, David R Wilson, John Choi, Mahita Varanasi, Katie Sanders, Johan Karlsson, Michael Lim, Jordan J Green
ABSTRACT
Efficient cytosolic protein delivery is necessary to fully realize the potential of protein therapeutics. Current methods of protein delivery often suffer from low serum tolerance and limited in vivo efficacy. Here, we report the synthesis and validation of a previously unreported class of carboxylated branched poly(β-amino ester)s that can self-assemble into nanoparticles for efficient intracellular delivery of a variety of different proteins. In vitro, nanoparticles enabled rapid cellular uptake, efficient endosomal escape, and functional cytosolic protein release into cells in media containing 10% serum. Moreover, nanoparticles encapsulating CRISPR-Cas9 ribonucleoproteins (RNPs) induced robust levels of gene knock-in (4%) and gene knockout (>75%) in several cell types. A single intracranial administration of nanoparticles delivering a low RNP dose (3.5 pmol) induced robust gene editing in mice bearing engineered orthotopic murine glioma tumors. This self-assembled polymeric nanocarrier system enables a versatile protein delivery and gene editing platform for biological research and therapeutic applications.
Product Description
Supelco
γ-Aminobutyric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
8-Aminooctanoic acid, 99%
TOYOPEARL® HW-55S Bulk Media, phase hydroxylated methacrylic polymer, bottle of 1 L, 30 μm particle size