MilliporeSigma
  • Screening of the Suzuki Cross-Coupling Reaction Using Desorption Electrospray Ionization in High-Throughput and in Leidenfrost Droplet Experiments.
Screening of the Suzuki Cross-Coupling Reaction Using Desorption Electrospray Ionization in High-Throughput and in Leidenfrost Droplet Experiments.
Journal of the American Society for Mass Spectrometry (2019-08-09)
Patrick W Fedick, Kiran Iyer, Zhenwei Wei, Larisa Avramova, Grace O Capek, R Graham Cooks
ABSTRACT
Suzuki cross-coupling is a widely performed reaction, typically using metal catalysts under heated conditions. Acceleration of the Suzuki cross-coupling reaction has been previously explored in microdroplets using desorption electrospray ionization mass spectrometry (DESI-MS). Building upon previous work, presented here is the use of a high-throughput DESI-MS screening system to identify optimal reaction conditions. Multiple reagents, bases, and stoichiometries were screened using the automated system at rates that approach 10,000 reaction mixture systems per hour. The DESI-MS system utilizes reaction acceleration in microdroplets to allow rapid screening. The results of screening of an array of reaction mixtures using this technique are presented as product ion images via standard MS imaging software, facilitating quick readout. Instructive comparisons are provided with another method of generating droplets for reaction acceleration-the Leidenfrost technique. Acceleration factors greater than 200 were measured for brominated substrates, paralleling the DESI-MS results. Acceleration factors dropped to near unity with highly substituted pyridines, attributable to a steric effect. The reaction proceeded in the absence of a base in Leidenfrost droplets although no product formation was seen without base in the bulk or in the DESI-MS screening experiments. These differences between Leidenfrost chemistry and the bulk and in droplets formed in high-throughput DESI are tentatively attributed to extremes of pH associated with the surfaces of Leidenfrost droplets.
Sigma-Aldrich
4-Hydroxyphenylboronic acid, ≥95.0%
Sigma-Aldrich
3-Iodopyridine, 98%
Sigma-Aldrich
3-Bromoquinoline, 98%
Sigma-Aldrich
3-Bromo-2,6-dimethylpyridine, 97%
Sigma-Aldrich
3-Bromopyridine, 99%