MilliporeSigma
  • Validation of Calibration Parameters for Trapped Ion Mobility Spectrometry.
Validation of Calibration Parameters for Trapped Ion Mobility Spectrometry.
Journal of the American Society for Mass Spectrometry (2019-08-09)
Cameron N Naylor, Tobias Reinecke, Mark E Ridgeway, Melvin A Park, Brian H Clowers
ABSTRACT
Using contemporary theory for ion mobility spectrometry (IMS), gas-phase ion mobilities within a trapped ion mobility-mass spectrometer (TIMS) are not easily deduced using first principle equations due to non-linear pressure changes and consequently variations in E/N. It is for this reason that prior literature values have traditionally been used for TIMS calibration. Additionally, given that verified mobility standards currently do not exist and the that the exact conditions used to measure reported literature values may not always represent the environment within the TIMS, a direct approach to validating the behavior of the TIMS system is warranted. A calibration procedure is presented where an ambient pressure, ambient temperature, two-gate, printed circuit board drift-tube IMS (PCBIMS) is coupled to the front of a TIMS allowing reduced mobilities to be directly measured on the same instrument as the TIMS. These measured mobilities were used to evaluate the TIMS calibration procedure which correlates reduced mobility and TIMS elution voltages with literature values. When using the measured PCBIMS-reduced mobilities of tetraalkyl ammonium salts and tune mix for TIMS calibration of the alkyltrimethyl ammonium salts, the percent error is less than 1% as compared with using the reported literature K0 values where the percent error approaches 5%. This method provides a way to obtain accurate reference mobilities for ion mobility techniques that require a calibration step (i.e., TIMS and TWAVE).
Sigma-Aldrich
Trimethyloctadecylammonium bromide, 98%
Sigma-Aldrich
Dodecyltrimethylammonium chloride, purum, ≥98.0% anhydrous basis (AT)
Sigma-Aldrich
Decyltrimethylammonium bromide, ≥98.0% (NT)
Sigma-Aldrich
Tetradodecylammonium bromide, ≥99.0% (AT)
Sigma-Aldrich
Benzyltriethylammonium chloride, 99%
Sigma-Aldrich
Trimethyl-tetradecylammonium chloride, ≥98.0% (AT)