MilliporeSigma
  • The response of phyllodes tumor of the breast to anticancer therapy: An in vitro and ex vivo study.
The response of phyllodes tumor of the breast to anticancer therapy: An in vitro and ex vivo study.
Oncology letters (2019-10-16)
Alicja Urbaniak, Fariba Jousheghany, Youzhong Yuan, Sergio Piña-Oviedo, Adam Huczyński, Magdalena Delgado, Thomas Kieber-Emmons, Behjatolah Monzavi-Karbassi, Timothy C Chambers
ABSTRACT
Phyllodes tumors of the breast (PTB) are uncommon stromal-epithelial neoplasms, with the main recommended treatment being surgical removal. However, even with adequate resection, the risk of recurrence in the malignant form remains as high as 40%, and there is no recognized consensus on the most effective drugs for PTB. In the present study, an ex vivo model of malignant phyllodes and derived primary cell cultures were used to evaluate the effectiveness of a panel of different drugs, including the Bcl-2/Bcl-xL inhibitor ABT-263, salinomycin (SAL), doxorubicin (DOX), paclitaxel (TAX), vincristine (VCR), colchicine (COL) and cisplatin (CIS). ABT-263, SAL and DOX were highly effective towards phyllodes spindle cells when assessed in the ex vivo model, contributing to ~98% tumor cell death. Furthermore, ABT-263 was highly selective for tumor cells in this system, and exhibited little toxic effect on adjacent normal epithelial cells. Furthermore, consistent with findings in the ex vivo model, ABT-263 was significantly less toxic towards MCF 10A non-tumorigenic breast epithelial cells compared with SAL and DOX. A conditional reprogramming strategy was subsequently used, involving Rho kinase inhibition, to successfully generate primary phyllodes tumor cells that could be cultured for several passages. The primary cells were sensitive to DOX with an IC50 of 0.40±0.07 µM in a standard viability assay and the preliminary results were obtained indicating sensitivity to ABT-263 and SAL. The present study demonstrated the feasibility of using explants and primary cells for drug discovery, selectively targeting PTB cells.
Product Description
SAFC
Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12, liquid, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Supelco
Doxorubicin Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Gentamicin solution, 50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture