MilliporeSigma
Tox4 modulates cell fate reprogramming.
Journal of cell science (2019-09-15)
Lotte Vanheer, Juan Song, Natalie De Geest, Adrian Janiszewski, Irene Talon, Caterina Provenzano, Taeho Oh, Joel Chappell, Vincent Pasque
ABSTRACT
Reprogramming to induced pluripotency induces the switch of somatic cell identity to induced pluripotent stem cells (iPSCs). However, the mediators and mechanisms of reprogramming remain largely unclear. To elucidate the mediators and mechanisms of reprogramming, we used a siRNA-mediated knockdown approach for selected candidate genes during the conversion of somatic cells into iPSCs. We identified Tox4 as a novel factor that modulates cell fate through an assay that determined the efficiency of iPSC reprogramming. We found that Tox4 is needed early in reprogramming to efficiently generate early reprogramming intermediates, irrespective of the reprogramming conditions used. Tox4 enables proper exogenous reprogramming factor expression, and the closing and opening of putative somatic and pluripotency enhancers early during reprogramming, respectively. We show that the TOX4 protein assembles into a high molecular form. Moreover, Tox4 is also required for the efficient conversion of fibroblasts towards the neuronal fate, suggesting a broader role of Tox4 in modulating cell fate. Our study reveals Tox4 as a novel transcriptional modulator of cell fate that mediates reprogramming from the somatic state to the pluripotent and neuronal fate.This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
Anti-TOX4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
DAPI, for nucleic acid staining