MilliporeSigma
  • Ahi1 promotes Arl13b ciliary recruitment, regulates Arl13b stability and is required for normal cell migration.
Ahi1 promotes Arl13b ciliary recruitment, regulates Arl13b stability and is required for normal cell migration.
Journal of cell science (2019-08-09)
Jesús Muñoz-Estrada, Russell J Ferland
ABSTRACT
Mutations in the Abelson-helper integration site 1 (AHI1) gene are associated with neurological/neuropsychiatric disorders, and cause the neurodevelopmental ciliopathy Joubert syndrome (JBTS). Here, we show that deletion of the transition zone (TZ) protein Ahi1 in mouse embryonic fibroblasts (MEFs) has a small effect on cilia formation. However, Ahi1 loss in these cells results in: (1) reduced localization of the JBTS-associated protein Arl13b to the ciliary membrane, (2) decreased sonic hedgehog signaling, (3) and an abnormally elongated ciliary axoneme accompanied by an increase in ciliary IFT88 concentrations. While no changes in Arl13b levels are detected in crude cell membrane extracts, loss of Ahi1 significantly reduced the level of non-membrane-associated Arl13b and its stability via the proteasome pathway. Exogenous expression of Ahi1-GFP in Ahi1-/- MEFs restored ciliary length, increased ciliary recruitment of Arl13b and augmented Arl13b stability. Finally, Ahi1-/- MEFs displayed defects in cell motility and Pdgfr-α-dependent migration. Overall, our findings support molecular mechanisms underlying JBTS etiology that involve: (1) disruptions at the TZ resulting in defects of membrane- and non-membrane-associated proteins to localize to primary cilia, and (2) defective cell migration.This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
4-Ethylmorpholine, ≥97.0% (GC)
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid