MilliporeSigma
  • The role of APC-mediated actin assembly in microtubule capture and focal adhesion turnover.
The role of APC-mediated actin assembly in microtubule capture and focal adhesion turnover.
The Journal of cell biology (2019-09-01)
M Angeles Juanes, Daniel Isnardon, Ali Badache, Sophie Brasselet, Manos Mavrakis, Bruce L Goode
ABSTRACT
Focal adhesion (FA) turnover depends on microtubules and actin. Microtubule ends are captured at FAs, where they induce rapid FA disassembly. However, actin's roles are less clear. Here, we use polarization-resolved microscopy, FRAP, live cell imaging, and a mutant of Adenomatous polyposis coli (APC-m4) defective in actin nucleation to investigate the role of actin assembly in FA turnover. We show that APC-mediated actin assembly is critical for maintaining normal F-actin levels, organization, and dynamics at FAs, along with organization of FA components. In WT cells, microtubules are captured repeatedly at FAs as they mature, but once a FA reaches peak maturity, the next microtubule capture event leads to delivery of an autophagosome, triggering FA disassembly. In APC-m4 cells, microtubule capture frequency and duration are altered, and there are long delays between autophagosome delivery and FA disassembly. Thus, APC-mediated actin assembly is required for normal feedback between microtubules and FAs, and maintaining FAs in a state "primed" for microtubule-induced turnover.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Anti-phospho-Src family (Tyr416) Antibody, clone 2N8, rabbit monoclonal, culture supernatant, clone 2N8, Upstate®
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Bafilomycin A1 Ready Made Solution, 0.16 mM in DMSO, from Streptomyces griseus