MilliporeSigma
  • Biophysical characterization of the insertion of two potent antimicrobial peptides-Pin2 and its variant Pin2[GVG] in biological model membranes.
Biophysical characterization of the insertion of two potent antimicrobial peptides-Pin2 and its variant Pin2[GVG] in biological model membranes.
Biochimica et biophysica acta. Biomembranes (2019-11-05)
Brandt Bertrand, Sathishkumar Munusamy, José-Francisco Espinosa-Romero, Gerardo Corzo, Iván Arenas Sosa, Arturo Galván-Hernández, Iván Ortega-Blake, Pablo Luis Hernández-Adame, Jaime Ruiz-García, José-Luis Velasco-Bolom, Ramón Garduño-Juárez, Carlos Munoz-Garay
ABSTRACT
The aim of this study was to investigate the factors that govern the activity and selectivity of two potent antimicrobial peptides (AMPs) using lipid membrane models of bacterial, erythrocyte and fungal cells. These models were used in calcein liposome leakage experiments to explore peptide efficiency. The AMPs (Pin2 and its variant Pin2[GVG]) showed highest affinity towards the bacterial models in the nanomolar range, followed by the erythrocyte and fungal systems. The presence of sterols modulated the variant's selectivity, while the wild type was unaffected. Liposome leakage experiments with Fluorescein Isothiocyanate-dextran (FITC)-dextran conjugates indicated that pore size depended on peptide concentration. Dynamic Light Scattering revealed peptide aggregation in aqueous solution, and that aggregate size was related to activity. The interacting peptides did not alter liposome size, suggesting pore forming activity rather than detergent activity. Atomic Force Microscopy showed differential membrane absorption, being greater in the bacterial model compared to the mammalian model, and pore-like defects were observed. Electrophysiological assays with the Tip-Dip Patch Clamp method provided evidence of changes in the electrical resistance of the membrane. Membrane potential experiments showed that liposomes were also depolarized in the presence of the peptides. Both peptides increased the Laurdan Generalized Polarization of the bacterial model indicating increased viscosity, on the contrary, no effect was observed with the erythrocyte and the fungal models. Peptide membrane insertion and pore formation was corroborated with Langmuir Pressure-Area isotherms and Brewster Angle Microscopy. Finally, molecular dynamics simulations were used to get an insight into the molecular mechanism of action.
Product Description
Avanti
Cholesterol (ovine), Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PG, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PG, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Sigma-Aldrich
6-Dodecanoyl-N,N-dimethyl-2-naphthylamine, ≥97.0% (HPLC), suitable for fluorescence
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Avanti
Sphingomyelin (d18:1/12:0), Avanti Research - A Croda Brand
Avanti
16:0-18:1 PE, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PE, Avanti Research - A Croda Brand