MilliporeSigma
  • A nutritional memory effect counteracts benefits of dietary restriction in old mice.
A nutritional memory effect counteracts benefits of dietary restriction in old mice.
Nature metabolism (2019-11-20)
Oliver Hahn, Lisa F Drews, An Nguyen, Takashi Tatsuta, Lisonia Gkioni, Oliver Hendrich, Qifeng Zhang, Thomas Langer, Scott Pletcher, Michael J O Wakelam, Andreas Beyer, Sebastian Grönke, Linda Partridge
ABSTRACT
Dietary restriction (DR) during adulthood can greatly extend lifespan and improve metabolic health in diverse species. However, whether DR in mammals is still effective when applied for the first time at old age remains elusive. Here, we report results of a late-life DR switch experiment employing 800 mice, in which 24 months old female mice were switched from ad libitum (AL) to DR or vice versa. Strikingly, the switch from DR-to-AL acutely increases mortality, whereas the switch from AL-to-DR causes only a weak and gradual increase in survival, suggesting a memory of earlier nutrition. RNA-seq profiling in liver, brown (BAT) and white adipose tissue (WAT) demonstrate a largely refractory transcriptional and metabolic response to DR after AL feeding in fat tissue, particularly in WAT, and a proinflammatory signature in aged preadipocytes, which is prevented by chronic DR feeding. Our results provide evidence for a nutritional memory as a limiting factor for DR-induced longevity and metabolic remodeling of WAT in mammals.
Product Description
Avanti
18:1-12:0 NBD PG, Avanti Research - A Croda Brand 810166P, powder
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial