MilliporeSigma
  • In vitro skin penetration enhancement techniques: A combined approach of ethosomes and microneedles.
In vitro skin penetration enhancement techniques: A combined approach of ethosomes and microneedles.
International journal of pharmaceutics (2019-11-13)
C Bellefroid, A Lechanteur, B Evrard, D Mottet, F Debacq-Chainiaux, G Piel
ABSTRACT
Dermal administration of different macromolecules, such as nucleic acids, remains a real challenge because of the difficulty of crossing the main skin barrier, the stratum corneum (SC). To overcome this barrier, the use of deformable lipid-based nanovectors were developed to increase topical penetration through the SC and to promote the intercellular delivery of drugs. The purpose of this study is to compare the skin penetration of different liposome formulations according to their composition. In vitro and ex vivo experiments using Franz diffusion cells were performed to highlight the effect of (i) lipid charge, (ii) edge activators (EA) and (iii) ethanol on the diffusion properties of nanovectors. We showed that all formulations were not able to cross the SC. However, on a tape stripped skin, we showed that cationic formulations containing an EA and ethanol improved the skin penetration. The use of microneedles was considered to bypass the SC. We have shown that sodium cholate and ethanol were necessary to ensure an appropriate diffusion of liposomes into the dermis when applied by means of microneedles. This could be a promising approach to further deliver efficiently macromolecules such as genes into the skin.
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand
Avanti
16:0-12:0 NBD PC, Avanti Research - A Croda Brand 810131C
Avanti
16:0-12:0 NBD PC, Avanti Research - A Croda Brand 810131P, powder
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PG, Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PG, Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder