MilliporeSigma
  • Calcium Influx through Plasma-Membrane Nanoruptures Drives Axon Degeneration in a Model of Multiple Sclerosis.
Calcium Influx through Plasma-Membrane Nanoruptures Drives Axon Degeneration in a Model of Multiple Sclerosis.
Neuron (2019-01-29)
Maarten E Witte, Adrian-Minh Schumacher, Christoph F Mahler, Jan P Bewersdorf, Jonas Lehmitz, Alexander Scheiter, Paula Sánchez, Philip R Williams, Oliver Griesbeck, Ronald Naumann, Thomas Misgeld, Martin Kerschensteiner
ABSTRACT
Axon loss determines persistent disability in multiple sclerosis patients. Here, we use in vivo calcium imaging in a multiple sclerosis model to show that cytoplasmic calcium levels determine the choice between axon loss and survival. We rule out the endoplasmic reticulum, glutamate excitotoxicity, and the reversal of the sodium-calcium exchanger as sources of intra-axonal calcium accumulation and instead identify nanoscale ruptures of the axonal plasma membrane as the critical path of calcium entry.
Sigma-Aldrich
Kainic acid monohydrate, ≥99% (TLC)
Sigma-Aldrich
Caffeine, powder, ReagentPlus®