MilliporeSigma
ArhGEF37 assists dynamin 2 during clathrin-mediated endocytosis.
Journal of cell science (2019-03-31)
Abhiyan Viplav, Tanumoy Saha, Jan Huertas, Philipp Selenschik, Mirsana P Ebrahimkutty, David Grill, Julia Lehrich, Andreas Hentschel, Monika Biasizzo, Simone Mengoni, Robert Ahrends, Volker Gerke, Vlad Cojocaru, Jürgen Klingauf, Milos Galic
ABSTRACT
Clathrin-mediated endocytosis (CME) engages over 30 proteins to secure efficient cargo and membrane uptake. While the function of most core CME components is well established, auxiliary mechanisms crucial for fine-tuning and adaptation remain largely elusive. In this study, we identify ArhGEF37, a currently uncharacterized protein, as a constituent of CME. Structure prediction together with quantitative cellular and biochemical studies present a unique BAR domain and PI(4,5)P2-dependent protein-membrane interactions. Functional characterization yields accumulation of ArhGEF37 at dynamin 2-rich late endocytic sites and increased endocytosis rates in the presence of ArhGEF37. Together, these results introduce ArhGEF37 as a regulatory protein involved in endocytosis.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in glass vials
Sigma-Aldrich
Dynasore hydrate
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Roche
KAPA SYBR® FAST, suitable for qPCR, 2 ×, Bio-Rad iCycler®
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)