MilliporeSigma
  • Loss of MCU prevents mitochondrial fusion in G1-S phase and blocks cell cycle progression and proliferation.
Loss of MCU prevents mitochondrial fusion in G1-S phase and blocks cell cycle progression and proliferation.
Science signaling (2019-05-02)
Olha M Koval, Emily K Nguyen, Velarchana Santhana, Trevor P Fidler, Sara C Sebag, Tyler P Rasmussen, Dylan J Mittauer, Stefan Strack, Prabhat C Goswami, E Dale Abel, Isabella M Grumbach
ABSTRACT
The role of the mitochondrial Ca2+ uniporter (MCU) in physiologic cell proliferation remains to be defined. Here, we demonstrated that the MCU was required to match mitochondrial function to metabolic demands during the cell cycle. During the G1-S transition (the cycle phase with the highest mitochondrial ATP output), mitochondrial fusion, oxygen consumption, and Ca2+ uptake increased in wild-type cells but not in cells lacking MCU. In proliferating wild-type control cells, the addition of the growth factors promoted the activation of the Ca2+/calmodulin-dependent kinase II (CaMKII) and the phosphorylation of the mitochondrial fission factor Drp1 at Ser616 The lack of the MCU was associated with baseline activation of CaMKII, mitochondrial fragmentation due to increased Drp1 phosphorylation, and impaired mitochondrial respiration and glycolysis. The mitochondrial fission/fusion ratio and proliferation in MCU-deficient cells recovered after MCU restoration or inhibition of mitochondrial fragmentation or of CaMKII in the cytosol. Our data highlight a key function for the MCU in mitochondrial adaptation to the metabolic demands during cell cycle progression. Cytosolic CaMKII and the MCU participate in a regulatory circuit, whereby mitochondrial Ca2+ uptake affects cell proliferation through Drp1.
Sigma-Aldrich
Anti-CaM Kinase II Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
KN-93, ≥98% (HPLC)
Sigma-Aldrich
Aphidicolin, Ready Made Solution, from Nigrospora sphaerica
Sigma-Aldrich
Anti-MICU1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SLC24A6 antibody produced in rabbit, affinity isolated antibody
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests
Sigma-Aldrich
Ru360, Ru360, is a cell-permeable oxygen-bridged dinuclear ruthenium amine complex. Binds to mitochondria with high affinity (Kd = 340 pM) and blocks Ca2+ uptake into mitochondria in vitro (IC₅₀ = 184 pM).
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Caffeine, powder, ReagentPlus®