MilliporeSigma
  • Loss of Hepatic Oscillatory Fed microRNAs Abrogates Refed Transition and Causes Liver Dysfunctions.
Loss of Hepatic Oscillatory Fed microRNAs Abrogates Refed Transition and Causes Liver Dysfunctions.
Cell reports (2019-02-21)
Babukrishna Maniyadath, Tandrika Chattopadhyay, Srikant Verma, Sujata Kumari, Prineeta Kulkarni, Kushal Banerjee, Asmitha Lazarus, Saurabh S Kokane, Trupti Shetty, Krishanpal Anamika, Ullas Kolthur-Seetharam
ABSTRACT
Inability to mediate fed-fast transitions in the liver is known to cause metabolic dysfunctions and diseases. Intuitively, a failure to inhibit futile translation of state-specific transcripts during fed-fast cycles would abrogate dynamic physiological transitions. Here, we have discovered hepatic fed microRNAs that target fasting-induced genes and are essential for a refed transition. Our findings highlight the role of these fed microRNAs in orchestrating system-level control over liver physiology and whole-body energetics. By targeting SIRT1, PGC1α, and their downstream genes, fed microRNAs regulate metabolic and mitochondrial pathways. MicroRNA expression, processing, and RISC loading oscillate during these cycles and possibly constitute an anticipatory mechanism. Fed-microRNA oscillations are deregulated during aging. Scavenging of hepatic fed microRNAs causes uncontrolled gluconeogenesis and failure in the catabolic-to-anabolic switching upon feeding, which are hallmarks of metabolic diseases. Besides identifying mechanisms that enable efficient physiological toggling, our study highlights fed microRNAs as candidate therapeutic targets.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Anti-PGC-1α Mouse mAb (4C1.3), liquid, clone 4C1.3, Calbiochem®
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder