MilliporeSigma
  • MYC Recruits SPT5 to RNA Polymerase II to Promote Processive Transcription Elongation.
MYC Recruits SPT5 to RNA Polymerase II to Promote Processive Transcription Elongation.
Molecular cell (2019-04-01)
Apoorva Baluapuri, Julia Hofstetter, Nevenka Dudvarski Stankovic, Theresa Endres, Pranjali Bhandare, Seychelle Monique Vos, Bikash Adhikari, Jessica Denise Schwarz, Ashwin Narain, Markus Vogt, Shuang-Yan Wang, Robert Düster, Lisa Anna Jung, Jens Thorsten Vanselow, Armin Wiegering, Matthias Geyer, Hans Michael Maric, Peter Gallant, Susanne Walz, Andreas Schlosser, Patrick Cramer, Martin Eilers, Elmar Wolf
ABSTRACT
The MYC oncoprotein binds to promoter-proximal regions of virtually all transcribed genes and enhances RNA polymerase II (Pol II) function, but its precise mode of action is poorly understood. Using mass spectrometry of both MYC and Pol II complexes, we show here that MYC controls the assembly of Pol II with a small set of transcription elongation factors that includes SPT5, a subunit of the elongation factor DSIF. MYC directly binds SPT5, recruits SPT5 to promoters, and enables the CDK7-dependent transfer of SPT5 onto Pol II. Consistent with known functions of SPT5, MYC is required for fast and processive transcription elongation. Intriguingly, the high levels of MYC that are expressed in tumors sequester SPT5 into non-functional complexes, thereby decreasing the expression of growth-suppressive genes. Altogether, these results argue that MYC controls the productive assembly of processive Pol II elongation complexes and provide insight into how oncogenic levels of MYC permit uncontrolled cellular growth.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-GST Antibody, Cytiva 27-4577-01