MilliporeSigma
  • A Site of Vulnerability on the Influenza Virus Hemagglutinin Head Domain Trimer Interface.
A Site of Vulnerability on the Influenza Virus Hemagglutinin Head Domain Trimer Interface.
Cell (2019-05-18)
Sandhya Bangaru, Shanshan Lang, Michael Schotsaert, Hillary A Vanderven, Xueyong Zhu, Nurgun Kose, Robin Bombardi, Jessica A Finn, Stephen J Kent, Pavlo Gilchuk, Iuliia Gilchuk, Hannah L Turner, Adolfo García-Sastre, Sheng Li, Andrew B Ward, Ian A Wilson, James E Crowe
ABSTRACT
Here, we describe the discovery of a naturally occurring human antibody (Ab), FluA-20, that recognizes a new site of vulnerability on the hemagglutinin (HA) head domain and reacts with most influenza A viruses. Structural characterization of FluA-20 with H1 and H3 head domains revealed a novel epitope in the HA trimer interface, suggesting previously unrecognized dynamic features of the trimeric HA protein. The critical HA residues recognized by FluA-20 remain conserved across most subtypes of influenza A viruses, which explains the Ab's extraordinary breadth. The Ab rapidly disrupted the integrity of HA protein trimers, inhibited cell-to-cell spread of virus in culture, and protected mice against challenge with viruses of H1N1, H3N2, H5N1, or H7N9 subtypes when used as prophylaxis or therapy. The FluA-20 Ab has uncovered an exceedingly conserved protective determinant in the influenza HA head domain trimer interface that is an unexpected new target for anti-influenza therapeutics and vaccines.
Sigma-Aldrich
Brefeldin A, BioXtra, ≥99% (HPLC and TLC)