MilliporeSigma
  • Targeting TMEM176B Enhances Antitumor Immunity and Augments the Efficacy of Immune Checkpoint Blockers by Unleashing Inflammasome Activation.
Targeting TMEM176B Enhances Antitumor Immunity and Augments the Efficacy of Immune Checkpoint Blockers by Unleashing Inflammasome Activation.
Cancer cell (2019-05-16)
Mercedes Segovia, Sofia Russo, Mathias Jeldres, Yamil D Mahmoud, Valentina Perez, Maite Duhalde, Pierre Charnet, Matthieu Rousset, Sabina Victoria, Florencia Veigas, Cédric Louvet, Bernard Vanhove, R Andrés Floto, Ignacio Anegon, Maria Cristina Cuturi, M Romina Girotti, Gabriel A Rabinovich, Marcelo Hill
ABSTRACT
Although immune checkpoint blockers have yielded significant clinical benefits in patients with different malignancies, the efficacy of these therapies is still limited. Here, we show that disruption of transmembrane protein 176B (TMEM176B) contributes to CD8+ T cell-mediated tumor growth inhibition by unleashing inflammasome activation. Lack of Tmem176b enhances the antitumor activity of anti-CTLA-4 antibodies through mechanisms involving caspase-1/IL-1β activation. Accordingly, patients responding to checkpoint blockade therapies display an activated inflammasome signature. Finally, we identify BayK8644 as a potent TMEM176B inhibitor that promotes CD8+ T cell-mediated tumor control and reinforces the antitumor activity of both anti-CTLA-4 and anti-PD-1 antibodies. Thus, pharmacologic de-repression of the inflammasome by targeting TMEM176B may enhance the therapeutic efficacy of immune checkpoint blockers.
Sigma-Aldrich
Hydroxychloroquine sulfate, ≥98% (HPLC), powder
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Iberiotoxin, recombinant from Mesobuthus tamulus, ≥98% (HPLC), recombinant, expressed in E. coli
Sigma-Aldrich
CGP-37157, ≥98% (HPLC), powder
Sigma-Aldrich
Clotrimazole
Sigma-Aldrich
Nilvadipine, ≥98% (HPLC)