MilliporeSigma
  • Rhodium-catalyzed asymmetric conjugate addition of arylboronic acids to nitroalkenes using olefin-sulfoxide ligands.
Rhodium-catalyzed asymmetric conjugate addition of arylboronic acids to nitroalkenes using olefin-sulfoxide ligands.
The Journal of organic chemistry (2012-03-27)
Feng Xue, Dongping Wang, Xincheng Li, Boshun Wan
ABSTRACT
An efficient rhodium/olefin-sulfoxide catalyzed asymmetric conjugate addition of organoboronic acids to a variety of nitroalkenes has been developed, where 2-methoxy-1-naphthyl sulfinyl functionalized olefin ligands have shown to be highly effective and are applicable to a broad scope of aryl, alkyl, and heteroaryl nitroalkenes.
Sigma-Aldrich
4-Fluorophenylboronic acid, ≥95%
Sigma-Aldrich
4-Methoxyphenylboronic acid, ≥95.0%
Sigma-Aldrich
p-Tolylboronic acid, 97%