MilliporeSigma
  • The Transcriptional Regulator Sin3A Contributes to the Oncogenic Potential of STAT3.
The Transcriptional Regulator Sin3A Contributes to the Oncogenic Potential of STAT3.
Cancer research (2019-01-30)
Giovanni Gambi, Elisabetta Di Simone, Veronica Basso, Luisa Ricci, Rui Wang, Akanksha Verma, Olivier Elemento, Maurilio Ponzoni, Giorgio Inghirami, Laura Icardi, Anna Mondino
ABSTRACT
Epigenetic silencing of promoter and enhancer regions is a common phenomenon in malignant cells. The transcription factor STAT3 is aberrantly activated in several tumors, where its constitutive acetylation accounts for the transcriptional repression of a number of tumor suppressor genes (TSG) via molecular mechanisms that remain to be understood. Using nucleophosmin-anaplastic lymphoma kinase-positive (NPM-ALK+) anaplastic large-cell lymphoma (ALCL) as model system, we found in cells and patient-derived tumor xenografts that STAT3 is constitutively acetylated as a result of ALK activity. STAT3 acetylation relied on intact ALK-induced PI3K- and mTORC1-dependent signaling and was sensitive to resveratrol. Resveratrol lowered STAT3 acetylation, rescued TSG expression, and induced ALCL apoptotic cell death. STAT3 constitutively bound the Sin3A transcriptional repressor complex, and both STAT3 and Sin3A bound the promoter region of silenced TSG via a resveratrol-sensitive mechanism. Silencing SIN3A caused reexpression of TSG, induced ALCL apoptotic cell death in vitro, and hindered ALCL tumorigenic potential in vivo. A constitutive STAT3-Sin3A interaction was also found in breast adenocarcinoma cells and proved critical for TSG silencing and cell survival. Collectively, these results suggest that oncogene-driven STAT3 acetylation and its constitutive association with Sin3A represent novel and concomitant events contributing to STAT3 oncogenic potential. SIGNIFICANCE: This study delineates the transcriptional regulatory complex Sin3A as a mediator of STAT3 transcriptional repressor activity and identifies the STAT3/Sin3A axis as a druggable target to antagonize STAT3-addicted tumors. GRAPHICAL ABSTRACT: http://cancerres.aacrjournals.org/content/canres/79/12/3076/F1.large.jpg.See related commentary by Monteleone and Poli, p. 3031.
Sigma-Aldrich
IgG1 Isotype Control from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Crizotinib, ≥98% (HPLC)
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)