MilliporeSigma
  • Effect of poly-L-arginine on the nasal absorption of FITC-dextran of different molecular weights and recombinant human granulocyte colony-stimulating factor (rhG-CSF) in rats.
Effect of poly-L-arginine on the nasal absorption of FITC-dextran of different molecular weights and recombinant human granulocyte colony-stimulating factor (rhG-CSF) in rats.
International journal of pharmaceutics (2001-09-05)
M Miyamoto, H Natsume, I Satoh, K Ohtake, M Yamaguchi, D Kobayashi, K Sugibayashi, Y Morimoto
ABSTRACT
The effect of poly-L-arginine (poly-L-Arg) on the in vivo nasal absorption of FITC-dextrans with a mean molecular weight ranging from 4.3 to 167 kDa and recombinant human granulocyte colony-stimulating factor (rhG-CSF) in rats were studied. When FITC-dextrans were co-administered intranasally with 1.0 w/v% poly-L-Args of different molecular weight (MW, ca. 45.5 and 92 kDa, poly-L-Arg (50) and poly-L-Arg (100)), the bioavailability (F(infinity)) increased markedly compared with that after administration of FITC-dextran alone. However, the F(infinity) decreased exponentially with the increasing molecular weight of FITC-dextrans. There was no significant difference between the enhanced nasal absorption of FITC-dextrans achieved by the co-administration of poly-L-Arg (50) and poly-L-Arg (100). Moreover, the relationship between the F(infinity) and the molecular weight of FITC-dextrans indicated that the molecular weight of protein drugs, which exhibited efficient absorption with poly-L-Arg, was about 20 kDa, when the lower limit of bioavailability for developing a potent transnasal delivery system was assumed to be about 10%. Indeed, the nasal absorption of rhG-CSF, which has a molecular weight of 18.8 kDa, was also increased after co-administration of 1.0 w/v% poly-L-Arg (50) and the F(infinity) was about 11%. It seems likely that poly-L-Arg can be used to provide adequate nasal absorption of various protein drugs which have a molecular weight of about 20 kDa, thereby allowing the successful development of a variety of transnasal drug delivery systems.
Sigma-Aldrich
Poly-L-arginine hydrochloride, mol wt 5,000-15,000
Sigma-Aldrich
Poly-L-arginine hydrochloride, mol wt 15,000-70,000
Sigma-Aldrich
Poly-L-arginine hydrochloride, mol wt >70,000