MilliporeSigma
Alteration of the nuclear pore complex in Ca(2+)-mediated cell death.
Cell death and differentiation (2009-08-29)
D Bano, D Dinsdale, A Cabrera-Socorro, S Maida, N Lambacher, B McColl, E Ferrando-May, M O Hengartner, P Nicotera
ABSTRACT
Cell death requires coordinated intracellular signalling before disassembly of cell architecture by degradative enzymes. Although the death signalling cascades that involve the mitochondria, the ER and the plasma membrane have been extensively characterized, only a handful of studies have examined the functional and structural alterations of the nuclear pore complex (NPC) during neuronal death. Here, we show that during excitotoxic neuronal degeneration calpains redistributed across the nuclear envelope and mediated the degradation of NPC components causing altered permeability of the nuclear membrane. In primary dissociated neurons, simultaneous recording of cytosolic [Ca(2+)] and localization of fluorescent proteins showed that the onset of Ca(2+) overload signalled a progressive increase in the diffusion of small reporter molecules across the nuclear envelope. Later, calpain-mediated changes in nuclear pore permeability allowed accumulation of large proteins in the nucleus. Further, in a model of excitotoxic neuronal degeneration in Caenorhabditis elegans, we found similar nuclear changes and redistribution of fluorescent probes across the nuclear membrane in dying neurons. Our findings strongly suggest that increased leakiness of the nuclear barrier affects nucleocytoplasmic transport, alters the localization of proteins across the nuclear envelope and it is likely to be involved in Ca(2+)-dependent cell death, including ischemic neuronal demise.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid