MilliporeSigma
  • Tracking, Behavior and Fate of 58 Pesticides Originated from Hops during Beer Brewing.
Tracking, Behavior and Fate of 58 Pesticides Originated from Hops during Beer Brewing.
Journal of agricultural and food chemistry (2018-09-04)
Martin Dušek, Vladimíra Jandovská, Jana Olšovská
ABSTRACT
The study presents tracking of 58 pesticide residues associated with hops to estimate their carryover into brewed beer. The pesticides were spiked onto organic hops at a concentration of 15 mg/kg, and the wort was boiled with the artificially contaminated hops and fermented on a laboratory scale. Samples were collected during the whole brewing process and pesticide residues were extracted using a method known as QuEChERS (quick, easy, cheap, effective, rugged, and safe). An HPLC-HR-MS/MS method was developed and validated to identify and quantitate pesticide residues in treated hops, spent hops, hopped wort, green beer, and beer samples. Quantitation was achieved using standard addition with isotopically labeled standards. The carryover percentages into hopped wort and the percentages of decay reduction relative to the amount spiked on hops were calculated. The relationship between the partition coefficients n-octanol-water (log P values) and the residual ratios ( RW and RB) of a pesticide were evaluated to predict their behavior during hopping of wort and fermentation. Pesticides with a high log P values (>3.75) tended to remain in spent hops. The pesticides that have a low log P value up to approximately 3 could represent the demarcation lines of appreciable transfer rate of pesticides from hops to beer. Consequently, the pesticides were divided into three categories depending upon their fate during the brewing process. The most potential risk category represents a group involving the thermostable pesticides, such as azoxystrobin, boscalid, dimethomorph, flonicamid, imidacloprid, mandipropamid, myclobutanil, and thiamethoxam, which were transferred at high rates from the pesticide enriched hops into beer during the laboratory brewing trial. These results can be used as a guideline in the application of pesticides on hop plants that would reduce the level of pesticide residues in beer and their exposure in humans.
Product Description
Supelco
Azoxystrobin, PESTANAL®, analytical standard
Supelco
Fluopicolide, PESTANAL®, analytical standard
Supelco
Fludioxonil, PESTANAL®, analytical standard
Supelco
Fenpyroximate, PESTANAL®, analytical standard
Supelco
Oxadiazon, PESTANAL®, analytical standard
Supelco
Tebufenozide, PESTANAL®, analytical standard
Supelco
Clothianidin, PESTANAL®, analytical standard
Supelco
Dimethomorph, PESTANAL®, analytical standard
Supelco
Propamocarb, PESTANAL®, analytical standard
Supelco
Trifloxystrobin, PESTANAL®, analytical standard
Supelco
Thiamethoxam, PESTANAL®, analytical standard
Supelco
Tebuconazole, PESTANAL®, analytical standard
Supelco
Spirotetramat, PESTANAL®, analytical standard
Supelco
Spiroxamine, PESTANAL®, analytical standard
Supelco
Spirodiclofen, PESTANAL®, analytical standard
Supelco
Quinoxyfen, PESTANAL®, analytical standard
Supelco
Pirimicarb, PESTANAL®, analytical standard
Supelco
N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine, PESTANAL®, analytical standard
Supelco
Myclobutanil, PESTANAL®, analytical standard
Supelco
Metrafenone, PESTANAL®, analytical standard
Supelco
Indoxacarb, PESTANAL®, analytical standard
Supelco
Methoxyfenozide, PESTANAL®, analytical standard
Supelco
Mandipropamid, PESTANAL®, analytical standard
Supelco
Fluopyram, PESTANAL®, analytical standard
Supelco
Thiacloprid, PESTANAL®, analytical standard
Supelco
Cymoxanil, PESTANAL®, analytical standard
Supelco
Pyridaben, PESTANAL®, analytical standard
Supelco
Etoxazole, PESTANAL®, analytical standard
Supelco
Bupirimate, PESTANAL®, analytical standard
Supelco
Diflubenzuron, PESTANAL®, analytical standard