MilliporeSigma
  • E proteins sharpen neurogenesis by modulating proneural bHLH transcription factors' activity in an E-box-dependent manner.
E proteins sharpen neurogenesis by modulating proneural bHLH transcription factors' activity in an E-box-dependent manner.
eLife (2018-08-11)
Gwenvael Le Dréau, René Escalona, Raquel Fueyo, Antonio Herrera, Juan D Martínez, Susana Usieto, Anghara Menendez, Sebastian Pons, Marian A Martinez-Balbas, Elisa Marti
ABSTRACT
Class II HLH proteins heterodimerize with class I HLH/E proteins to regulate transcription. Here, we show that E proteins sharpen neurogenesis by adjusting the neurogenic strength of the distinct proneural proteins. We find that inhibiting BMP signaling or its target ID2 in the chick embryo spinal cord, impairs the neuronal production from progenitors expressing ATOH1/ASCL1, but less severely that from progenitors expressing NEUROG1/2/PTF1a. We show this context-dependent response to result from the differential modulation of proneural proteins' activity by E proteins. E proteins synergize with proneural proteins when acting on CAGSTG motifs, thereby facilitating the activity of ASCL1/ATOH1 which preferentially bind to such motifs. Conversely, E proteins restrict the neurogenic strength of NEUROG1/2 by directly inhibiting their preferential binding to CADATG motifs. Since we find this mechanism to be conserved in corticogenesis, we propose this differential co-operation of E proteins with proneural proteins as a novel though general feature of their mechanism of action.
Product Description
Rabbit IgG HRP Linked Whole Ab, Cytiva NA934-1ML
Sigma-Aldrich
Anti-Tubulin Antibody, beta, clone KMX-1, clone KMX-1, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-phospho-Histone H3 (pSer28) antibody produced in rat, ~0.5 mg/mL, clone HTA28, purified immunoglobulin, buffered aqueous solution
Millipore
Protein A Agarose, Protein A Agarose Beads for the purification of human, mouse & rabbit immunoglobins.