MilliporeSigma
  • Dual role of the RNA helicase DDX5 in post-transcriptional regulation of myelin basic protein in oligodendrocytes.
Dual role of the RNA helicase DDX5 in post-transcriptional regulation of myelin basic protein in oligodendrocytes.
Journal of cell science (2018-04-07)
Peter Hoch-Kraft, Robin White, Stefan Tenzer, Eva-Maria Krämer-Albers, Jacqueline Trotter, Constantin Gonsior
ABSTRACT
In the central nervous system, oligodendroglial expression of myelin basic protein (MBP) is crucial for the assembly and structure of the myelin sheath. MBP synthesis is tightly regulated in space and time, particularly at the post-transcriptional level. We have identified the DEAD-box RNA helicase DDX5 (also known as p68) in a complex with Mbp mRNA in oligodendroglial cells. Expression of DDX5 is highest in progenitor cells and immature oligodendrocytes, where it localizes to heterogeneous populations of cytoplasmic ribonucleoprotein (RNP) complexes associated with Mbp mRNA in the cell body and processes. Manipulation of the amount of DDX5 protein inversely affects the level of MBP. We present evidence that DDX5 is involved in post-transcriptional regulation of MBP protein synthesis, with implications for oligodendroglial development. In addition, knockdown of DDX5 results in an increased abundance of MBP isoforms containing exon 2 in immature oligodendrocytes, most likely by regulating alternative splicing of Mbp Our findings contribute to the understanding of the complex nature of MBP post-transcriptional control in immature oligodendrocytes where DDX5 appears to affect the abundance of MBP proteins via distinct but converging mechanisms.
Sigma-Aldrich
Anti-hnRNP-A2/B1 antibody, Mouse monoclonal, clone DP3B3, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human DDX5
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-c-Myc antibody produced in mouse, clone 9E10, ascites fluid
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid