MilliporeSigma
  • Single Live Cell Monitoring of Protein Turnover Reveals Intercellular Variability and Cell-Cycle Dependence of Degradation Rates.
Single Live Cell Monitoring of Protein Turnover Reveals Intercellular Variability and Cell-Cycle Dependence of Degradation Rates.
Molecular cell (2018-08-28)
Andrea Brigitta Alber, Eric Raphael Paquet, Martina Biserni, Felix Naef, David Michael Suter
ABSTRACT
Cells need to reliably control their proteome composition to maintain homeostasis and regulate growth. How protein synthesis and degradation interplay to control protein expression levels remains unclear. Here, we combined a tandem fluorescent timer and pulse-chase protein labeling to disentangle how protein synthesis and degradation control protein homeostasis in single live mouse embryonic stem cells. We discovered substantial cell-cycle dependence in protein synthesis rates and stabilization of a large number of proteins around cytokinesis. Protein degradation rates were highly variable between cells, co-varied within individual cells for different proteins, and were positively correlated with synthesis rates. This suggests variability in proteasome activity as an important source of global extrinsic noise in gene expression. Our approach paves the way toward understanding the complex interplay of synthesis and degradation processes in determining protein levels of individual mammalian cells.
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
GSK-3 Inhibitor XVI, GSK-3 Inhibitor XVI - CAS 252917-06-9, is a cell-permeable, potent, ATP-competitive, and highly selective GSK-3 inhibitor (IC₅₀ = 10 and 6.7 nM against GSK-3α and GSK-3β, respectively).
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Glasgow Minimum Essential Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture