MilliporeSigma
  • Cyclin D1 depletion interferes with oxidative balance and promotes cancer cell senescence.
Cyclin D1 depletion interferes with oxidative balance and promotes cancer cell senescence.
Journal of cell science (2018-06-09)
Phatthamon Laphanuwat, Pornlada Likasitwatanakul, Gunya Sittithumcharee, Araya Thaphaengphan, Nussara Chomanee, Orawan Suppramote, Nuttavadee Ketaroonrut, Komgrid Charngkaew, Eric W-F Lam, Seiji Okada, Uraiwan Panich, Somponnat Sampattavanich, Siwanon Jirawatnotai
ABSTRACT
Expression of cyclin D1 (CCND1) is required for cancer cell survival and proliferation. This is presumably due to the role of cyclin D1 in inactivation of the RB tumor suppressor. Here, we investigated the pro-survival function of cyclin D1 in a number of cancer cell lines. We found that cyclin D1 depletion facilitated cellular senescence in several cancer cell lines. Senescence triggered by cyclin D1 depletion was more extensive than that caused by the prolonged CDK4 inhibition. Intriguingly, the senescence caused by cyclin D1 depletion was independent of RB status of the cancer cell. We identified a build-up of intracellular reactive oxygen species in the cancer cells that underwent senescence upon depletion of cyclin D1 but not in those cells where CDK4 was inhibited. The higher ROS levels were responsible for the cell senescence, which was instigated by the p38-JNK-FOXO3a-p27 pathway. Therefore, expression of cyclin D1 prevents cancer cells from undergoing senescence, at least partially, by keeping the level of intracellular oxidative stress at a tolerable sub-lethal level. Depletion of cyclin D1 promotes the RB-independent pro-senescence pathway and the cancer cells then succumb to the endogenous oxidative stress levels.This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
Anti-FOXO3a/FKHRL1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
PD 0332991 isethionate, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human CDKN1B
Sigma-Aldrich
Anti-MnSOD Antibody, clone MnS-1, clone MnS-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit