MilliporeSigma
  • Hepatic oxidative stress in pigmented P23H rhodopsin transgenic rats with progressive retinal degeneration.
Hepatic oxidative stress in pigmented P23H rhodopsin transgenic rats with progressive retinal degeneration.
Free radical biology & medicine (2018-07-15)
Lorena Perdices, Lorena Fuentes-Broto, Francisco Segura, Neyla Ben Gdara, Ana Isabel Sánchez-Cano, Gema Insa, Elvira Orduna, Isabel Pinilla
ABSTRACT
Retinitis pigmentosa (RP) comprises a group of inherited retinal degenerative conditions characterized by primary degeneration of the rod photoreceptors. Increased oxidative damage is observed in the retina, aqueous humor, and plasma of RP animal models and patients. The hepatic oxidative status may also be affected in RP due to oxidative damage influencing soluble macromolecules exiting the retina or to alterations in the melanopsin system resulting in chronic circadian desynchronization that negatively alters the oxidative stress defense system. P23H rats were crossed with pigmented Long Evans rats to produce offspring exhibiting the clinical conditions of RP. We measured hepatic malondialdehyde and 4-hydroxyalkenal concentrations as oxidative stress markers; nitrite level as a total nitrosative damage marker; total antioxidant capacity; and the activities of catalase, superoxide dismutase (SOD), and glutathione S-transferase. Retinal visual function was assessed based on optomotor and electroretinogram responses. P23H transgenic rats exhibited diminished visual acuity, contrast sensitivity, and electroretinographic responses according to the level of retinal degeneration. P23H rats at 30 days of age already demonstrated only 47% of the hepatic total antioxidant capacity of wild-type animals. Hepatic catalase and SOD activities were also reduced in P23H rats after 120 days, but we detected no difference in glutathione S-transferase activity. P23H rats had increased hepatic oxidative and nitrosative damage markers. GSH/GSSG ratio showed a significant diminution in P23H rats at P120 compared to WT. We conclude that the liver is under increased oxidative stress in P23H rats. Further studies are required, however, to clarify the contribution of systemic oxidative damage to the pathogenesis of RP.
Sigma-Aldrich
Myoglobin from equine heart, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
1-Methyl-2-phenylindole, 99%
Sigma-Aldrich
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Sigma-Aldrich
Albumin from mouse serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Supelco
Bradford Reagent, for 0.1-1.4 mg/ml protein
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade III, ammonium sulfate suspension, ≥0.8 units/mg protein
Sigma-Aldrich
Glutathione Reductase from baker′s yeast (S. cerevisiae), ammonium sulfate suspension, 100-300 units/mg protein (biuret)
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
1,1,3,3-Tetramethoxypropane, 99%
Sigma-Aldrich
Triton X-100, laboratory grade