MilliporeSigma
  • RNA editing derived epitopes function as cancer antigens to elicit immune responses.
RNA editing derived epitopes function as cancer antigens to elicit immune responses.
Nature communications (2018-09-27)
Minying Zhang, Jens Fritsche, Jason Roszik, Leila J Williams, Xinxin Peng, Yulun Chiu, Chih-Chiang Tsou, Franziska Hoffgaard, Valentina Goldfinger, Oliver Schoor, Amjad Talukder, Marie A Forget, Cara Haymaker, Chantale Bernatchez, Leng Han, Yiu-Huen Tsang, Kathleen Kong, Xiaoyan Xu, Kenneth L Scott, Harpreet Singh-Jasuja, Greg Lizee, Han Liang, Toni Weinschenk, Gordon B Mills, Patrick Hwu
ABSTRACT
In addition to genomic mutations, RNA editing is another major mechanism creating sequence variations in proteins by introducing nucleotide changes in mRNA sequences. Deregulated RNA editing contributes to different types of human diseases, including cancers. Here we report that peptides generated as a consequence of RNA editing are indeed naturally presented by human leukocyte antigen (HLA) molecules. We provide evidence that effector CD8+ T cells specific for edited peptides derived from cyclin I are present in human tumours and attack tumour cells that are presenting these epitopes. We show that subpopulations of cancer patients have increased peptide levels and that levels of edited RNA correlate with peptide copy numbers. These findings demonstrate that RNA editing extends the classes of HLA presented self-antigens and that these antigens can be recognised by the immune system.
Sigma-Aldrich
Polybrene Infection / Transfection Reagent, A highly efficient method of gene transfer into mammalian cells leveraging infection with retroviral vectors.
Sigma-Aldrich
Anti-CCNI antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ExtrAvidin® –Alkaline Phosphatase, buffered aqueous solution