MilliporeSigma
Application of Solvent Parameters for Predicting Organogel Formation.
AAPS PharmSciTech (2018-05-31)
Beibei Hu, Wei Sun, Baixue Yang, Heran Li, Liuchenzi Zhou, Sanming Li
ABSTRACT
Solvents, accounting the majority of the organogel system, have a tremendous impact on the characteristics of gels. To date, there is a large variety of organogel systems; relatively few have been investigated in the field of structure-solvent correlation. Here, a series of solvent parameters were applied to explore the role of solvent effect on network forming and gel property, intending to build the connection between the precise solvent parameter and gel property. Among the solvent parameters, Kamlet-Taft Parameters and Hansen solubility parameters can distinguish specific types of intermolecular interactions, which could correlate solvent parameter with the gel property. From an analysis of the morphologies obtained from POM and SEM, the gelator structure has an impact on its self-assembly. For possible conformations, the gelators were investigated through XRD. The investigation of solvent-property relationship will provide a theoretical basis for controllable drug delivery implants.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
L-Alanine methyl ester hydrochloride, 99%
Sigma-Aldrich
1,3-Propylene sulfite, 99%