MilliporeSigma
  • p38-mediated phosphorylation at T367 induces EZH2 cytoplasmic localization to promote breast cancer metastasis.
p38-mediated phosphorylation at T367 induces EZH2 cytoplasmic localization to promote breast cancer metastasis.
Nature communications (2018-07-20)
Talha Anwar, Caroline Arellano-Garcia, James Ropa, Yu-Chih Chen, Hong Sun Kim, Euisik Yoon, Sierrah Grigsby, Venkatesha Basrur, Alexey I Nesvizhskii, Andrew Muntean, Maria E Gonzalez, Kelley M Kidwell, Zaneta Nikolovska-Coleska, Celina G Kleer
ABSTRACT
Overexpression of EZH2 in estrogen receptor negative (ER-) breast cancer promotes metastasis. EZH2 has been mainly studied as the catalytic component of the Polycomb Repressive Complex 2 (PRC2) that mediates gene repression by trimethylating histone H3 at lysine 27 (H3K27me3). However, how EZH2 drives metastasis despite the low H3K27me3 levels observed in ER- breast cancer is unknown. Here we show that in human invasive carcinomas and distant metastases, cytoplasmic EZH2 phosphorylated at T367 is significantly associated with ER- disease and low H3K27me3 levels. p38-mediated EZH2 phosphorylation at T367 promotes EZH2 cytoplasmic localization and potentiates EZH2 binding to vinculin and other cytoskeletal regulators of cell migration and invasion. Ectopic expression of a phospho-deficient T367A-EZH2 mutant is sufficient to inhibit EZH2 cytoplasmic expression, disrupt binding to cytoskeletal regulators, and reduce EZH2-mediated adhesion, migration, invasion, and development of spontaneous metastasis. These results point to a PRC2-independent non-canonical mechanism of EZH2 pro-metastatic function.
Sigma-Aldrich
LookOut® Mycoplasma PCR Detection Kit, Optimized for use with JumpStart Taq DNA Polymerase, D9307.
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Doxycycline Hydrochloride, Ready Made Solution, 100 mg/mL in DMSO, ≥95.0% (HPLC)
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
FIBRONECTIN FROM HUMAN PLASMA, liquid, 0.1% (Solution), BioReagent, suitable for cell culture