MilliporeSigma
  • Oxaloacetate induces apoptosis in HepG2 cells via inhibition of glycolysis.
Oxaloacetate induces apoptosis in HepG2 cells via inhibition of glycolysis.
Cancer medicine (2018-03-14)
Ye Kuang, Xiaoyun Han, Mu Xu, Qing Yang
ABSTRACT
Most cancer cells perform glycolysis despite having sufficient oxygen. The specific metabolic pathways of cancer cells have become the focus of cancer treatment. Recently, accumulating evidence indicates oxidative phosphorylation (OXPHOS) and glycolysis can be regulated with each other. Thus, we suggest that the glycolysis of cancer cells is inhibited by restoring or improving OXPHOS in cancer cells. In our study, we found that oxaloacetate (OA) induced apoptosis in HepG2 cells in vivo and in vitro. Meanwhile, we found that OA induced a decrease in the energy metabolism of HepG2 cells. Further results showed that the expression and activity of glycolytic enzymes were decreased with OA treatment. Conversely, the expression and activity of enzymes involved in the TCA cycle and OXPHOS were increased with OA treatment. The results indicate that OA can inhibit glycolysis through enhancement of OXPHOS. In addition, OA-mediated suppression of HIF1α, p-Akt, and c-myc led to a decrease in glycolysis level. Therefore, OA has the potential to be a novel anticancer drug.
Sigma-Aldrich
Phosphofructokinase (PFK) Activity Colorimetric Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
Bicinchoninic Acid Kit for Protein Determination, for 200-1000 μg/ml protein
Sigma-Aldrich
Hexokinase Colorimetric Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
IDH Activity Assay Kit, sufficient for 100 colorimetric tests