MilliporeSigma
  • Helicobacter pylori Infection Modulates Host Cell Metabolism through VacA-Dependent Inhibition of mTORC1.
Helicobacter pylori Infection Modulates Host Cell Metabolism through VacA-Dependent Inhibition of mTORC1.
Cell host & microbe (2018-05-11)
Ik-Jung Kim, Jeongmin Lee, Seung J Oh, Mee-Sup Yoon, Sung-Soo Jang, Robin L Holland, Michael L Reno, Mohammed N Hamad, Tatsuya Maeda, Hee Jung Chung, Jie Chen, Steven R Blanke
ABSTRACT
Helicobacter pylori (Hp) vacuolating cytotoxin (VacA) is a bacterial exotoxin that enters host cells and induces mitochondrial dysfunction. However, the extent to which VacA-dependent mitochondrial perturbations affect overall cellular metabolism is poorly understood. We report that VacA perturbations in mitochondria are linked to alterations in cellular amino acid homeostasis, which results in the inhibition of mammalian target of rapamycin complex 1 (mTORC1) and subsequent autophagy. mTORC1, which regulates cellular metabolism during nutrient stress, is inhibited during Hp infection by a VacA-dependent mechanism. This VacA-dependent inhibition of mTORC1 signaling is linked to the dissociation of mTORC1 from the lysosomal surface and results in activation of cellular autophagy through the Unc 51-like kinase 1 (Ulk1) complex. VacA intoxication results in reduced cellular amino acids, and bolstering amino acid pools prevents VacA-mediated mTORC1 inhibition. Overall, these studies support a model that Hp modulate host cell metabolism through the action of VacA at mitochondria.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Nutrient Mixture F-12 Ham, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Dispase® II, protease
Sigma-Aldrich
Bafilomycin A1 Ready Made Solution, 0.16 mM in DMSO, from Streptomyces griseus
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Uvomorulin/E-Cadherin antibody produced in rat, clone DECMA-1, ascites fluid, buffered aqueous solution
Sigma-Aldrich
Triton X-100, laboratory grade