MilliporeSigma
  • The O-GlcNAc transferase OGT interacts with and post-translationally modifies the transcription factor HOXA1.
The O-GlcNAc transferase OGT interacts with and post-translationally modifies the transcription factor HOXA1.
FEBS letters (2018-02-22)
Amandine Draime, Laure Bridoux, Magali Belpaire, Tamara Pringels, Hervé Degand, Pierre Morsomme, René Rezsohazy
ABSTRACT
HOXA1 belongs to the HOX family of transcription factors which are key regulators of animal development. Little is known about the molecular pathways controlling HOXA1. Recent data from our group revealed distinct partner proteins interacting with HOXA1. Among them, OGT is an O-linked N-acetylglucosamine (O-GlcNAc) transferase modifying a variety of proteins involved in different cellular processes including transcription. Here, we confirm OGT as a HOXA1 interactor, we characterise which domains of HOXA1 and OGT are required for the interaction, and we provide evidence that OGT post-translationally modifies HOXA1. Mass spectrometry experiments indeed reveal that HOXA1 can be phosphorylated on the AGGTVGSPQYIHHSY peptide and that upon OGT expression, the phosphate adduct is replaced by an O-GlcNAc group.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Anti-Glutathione-S-Transferase (GST) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Glutathione Sepharose 4B, Cytiva 17-0756-01, pack of 10 mL
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Monoclonal Anti-Glutathione-S-Transferase (GST) antibody produced in mouse, clone GST-2, ascites fluid
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt >300,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade