MilliporeSigma
  • Lethal (3) malignant brain tumor-like 2 (L3MBTL2) protein protects against kidney injury by inhibiting the DNA damage-p53-apoptosis pathway in renal tubular cells.
Lethal (3) malignant brain tumor-like 2 (L3MBTL2) protein protects against kidney injury by inhibiting the DNA damage-p53-apoptosis pathway in renal tubular cells.
Kidney international (2017-12-26)
Huihui Huang, Chunhua Xu, Yang Wang, Chenling Meng, Wenjing Liu, Yueshui Zhao, Xiao-Ru Huang, Wenxing You, Bo Feng, Zhi-Hua Zheng, Yu Huang, Hui-Yao Lan, Jinzhong Qin, Yin Xia
ABSTRACT
DNA damage contributes to renal tubular cell death during kidney injury, but how DNA damage in tubular cells is regulated is not fully understood. Lethal (3) malignant brain tumor-like 2 (L3MBTL2), a novel polycomb group protein, has been implicated in regulating chromatin architecture. However, the biological functions of L3MBTL2 are largely undefined. Here we found that L3MBTL2 was expressed in the nuclei of renal tubular epithelial cells in mice. Ablation of L3mbtl2 in renal tubular cells resulted in increases in nuclear DNA damage, p53 activation, apoptosis, tubular injury and kidney dysfunction after cisplatin treatment or unilateral ureteral obstruction. In vitro, inhibition of L3MBTL2 sequentially promoted histone γH2AX expression, p53 activation and apoptosis in cisplatin-treated mouse proximal tubular TKPTS cells. Inhibition of p53 activity attenuated the apoptosis induced by L3mbtl2 deficiency after cisplatin treatment both in vivo and in vitro. Intriguingly, unlike other polycomb proteins, L3MBTL2 was not recruited to DNA damage sites, but instead increased nuclear chromatin density and reduced initial DNA damage load. Thus, L3MBTL2 plays a protective role in kidney injury, in part by inhibiting the DNA damage-p53-apoptosis pathway.
Sigma-Aldrich
SITE Liquid Media Supplement (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
ApopTag Plus In Situ Apoptosis Fluorescein Detection Kit, The ApopTag Plus Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a fluorescein reporter molecule.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Roche
Nuclease S7, Micrococcal nuclease, from Staphylococcus aureus