MilliporeSigma
  • Structure of the BRAF-MEK complex reveals a kinase activity independent role for BRAF in MAPK signaling.
Structure of the BRAF-MEK complex reveals a kinase activity independent role for BRAF in MAPK signaling.
Cancer cell (2014-08-27)
Jacob R Haling, Jawahar Sudhamsu, Ivana Yen, Steve Sideris, Wendy Sandoval, Wilson Phung, Brandon J Bravo, Anthony M Giannetti, Ariana Peck, Alexandre Masselot, Tony Morales, Darin Smith, Barbara J Brandhuber, Sarah G Hymowitz, Shiva Malek
ABSTRACT
Numerous oncogenic mutations occur within the BRAF kinase domain (BRAF(KD)). Here we show that stable BRAF-MEK1 complexes are enriched in BRAF(WT) and KRAS mutant (MT) cells but not in BRAF(MT) cells. The crystal structure of the BRAF(KD) in a complex with MEK1 reveals a face-to-face dimer sensitive to MEK1 phosphorylation but insensitive to BRAF dimerization. Structure-guided studies reveal that oncogenic BRAF mutations function by bypassing the requirement for BRAF dimerization for activity or weakening the interaction with MEK1. Finally, we show that conformation-specific BRAF inhibitors can sequester a dormant BRAF-MEK1 complex resulting in pathway inhibition. Taken together, these findings reveal a regulatory role for BRAF in the MAPK pathway independent of its kinase activity but dependent on interaction with MEK.
Sigma-Aldrich
Anti-BRAF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Raf-1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-MEK1 Antibody, Upstate®, from rabbit