MilliporeSigma
  • SIRT4 Is a Lysine Deacylase that Controls Leucine Metabolism and Insulin Secretion.
SIRT4 Is a Lysine Deacylase that Controls Leucine Metabolism and Insulin Secretion.
Cell metabolism (2017-04-06)
Kristin A Anderson, Frank K Huynh, Kelsey Fisher-Wellman, J Darren Stuart, Brett S Peterson, Jonathan D Douros, Gregory R Wagner, J Will Thompson, Andreas S Madsen, Michelle F Green, R Michael Sivley, Olga R Ilkayeva, Robert D Stevens, Donald S Backos, John A Capra, Christian A Olsen, Jonathan E Campbell, Deborah M Muoio, Paul A Grimsrud, Matthew D Hirschey
ABSTRACT
Sirtuins are NAD+-dependent protein deacylases that regulate several aspects of metabolism and aging. In contrast to the other mammalian sirtuins, the primary enzymatic activity of mitochondrial sirtuin 4 (SIRT4) and its overall role in metabolic control have remained enigmatic. Using a combination of phylogenetics, structural biology, and enzymology, we show that SIRT4 removes three acyl moieties from lysine residues: methylglutaryl (MG)-, hydroxymethylglutaryl (HMG)-, and 3-methylglutaconyl (MGc)-lysine. The metabolites leading to these post-translational modifications are intermediates in leucine oxidation, and we show a primary role for SIRT4 in controlling this pathway in mice. Furthermore, we find that dysregulated leucine metabolism in SIRT4KO mice leads to elevated basal and stimulated insulin secretion, which progressively develops into glucose intolerance and insulin resistance. These findings identify a robust enzymatic activity for SIRT4, uncover a mechanism controlling branched-chain amino acid flux, and position SIRT4 as a crucial player maintaining insulin secretion and glucose homeostasis during aging.
Sigma-Aldrich
Anti-SIRT4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
Sigma-Aldrich
Alamethicin, Ready Made Solution from Trichoderma viride, 5 mg/mL in DMSO
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Histopaque®-1119, sterile-filtered, density: 1.119