MilliporeSigma
  • Colorectal cancer cells require glycogen synthase kinase-3β for sustaining mitosis via translocated promoter region (TPR)-dynein interaction.
Colorectal cancer cells require glycogen synthase kinase-3β for sustaining mitosis via translocated promoter region (TPR)-dynein interaction.
Oncotarget (2018-03-24)
Firli R P Dewi, Takahiro Domoto, Masaharu Hazawa, Akiko Kobayashi, Takayuki Douwaki, Toshinari Minamoto, Richard W Wong
ABSTRACT
Glycogen synthase kinase (GSK) 3β, which mediates fundamental cellular signaling pathways, has emerged as a potential therapeutic target for many types of cancer including colorectal cancer (CRC). During mitosis, GSK3β localizes in mitotic spindles and centrosomes, however its function is largely unknown. We previously demonstrated that translocated promoter region (TPR, a nuclear pore component) and dynein (a molecular motor) cooperatively contribute to mitotic spindle formation. Such knowledge encouraged us to investigate putative functional interactions among GSK3β, TPR, and dynein in the mitotic machinery of CRC cells. Here, we show that inhibition of GSK3β attenuated proliferation, induced cell cycle arrest at G2/M phase, and increased apoptosis of CRC cells. Morphologically, GSK3β inhibition disrupted chromosome segregation, mitotic spindle assembly, and centrosome maturation during mitosis, ultimately resulting in mitotic cell death. These changes in CRC cells were associated with decreased expression of TPR and dynein, as well as disruption of their functional colocalization with GSK3β in mitotic spindles and centrosomes. Clinically, we showed that TPR expression was increased in CRC databases and primary tumors of CRC patients. Furthermore, TPR expression in SW480 cells xenografted into mice was reduced following treatment with GSK3β inhibitors. Together, these results indicate that GSK3β sustains steady mitotic processes for proliferation of CRC cells via interaction with TPR and dynein, thereby suggesting that the therapeutic effect of GSK3β inhibition depends on induction of mitotic catastrophe in CRC cells.
Sigma-Aldrich
Anti-GSK3 Antibody, clone 4G-1E, clone 4G-1E, Upstate®, from mouse
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, serum, Upstate®
Sigma-Aldrich
Monoclonal Anti-Dynein (Intermediate Chain) antibody produced in mouse, clone 70.1, ascites fluid
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit